Learn More
Sarstedt Inc CONTAINER W/ SCREW CAP 200/CS
Supplier: Sarstedt Inc 75.562.400
Container with screw cap, closed urine transfer into a V-Monovette Urine, max. working volume: 100 ml, (xH): 57 x 76 mm, opening: 62 mm, transparent, graduated, material: PP, screw cap, cap: yellow, with safety label and transfer device, cap assembled, with safety label, sterile, 5 piece(s)/tube bag
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.