Learn More
Andwin Scientific CONTAINER HDPE 8OZ 250/CS
Supplier: Andwin Scientific 89009664
The HDPE Multipurpose Containers can be utilized for the storage of pathology, industrial, and environmental samples. This product is resistant to chemicals and breakage. The cap included snaps tightly and provides a secure fit. The container has a m