Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fischer Technical Company™ Contact Plates
Contact plates ideal for testing surfaces.
Supplier: Fischer Technical Company™ CM392RP15
Alert:
Prop 65: Cancer and Reproductive Harm.
Description
- Available in a variety of formulations
- For environmental monitoring of surfaces
- Steam sterilized
- Agar is slightly raised to make it easy to test surfaces
Specifications
Fungi | |
US with Globally Sourced Materials | |
Contact Plate | |
Fungi | |
Microbiological Testing | |
65 x 15 mm |
Contact Plate | |
SDA with Lecithin and Tween 80 | |
Sterile | |
Microbiological Tests | |
10 Plates |
Safety and Handling
ShelfLife : 90 Days
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction