Learn More
Enzo Life Sciences Congo Red (ultra pure) (100mg). CAS: 573-58-0
Supplier: Enzo Life Sciences ENZ52552
Congo Red, has been used in microscopy for staining elastic fibers and bacteria. Amyloid protein, an abnormal protein aggregate associated with various pathologies, is commonly detected in tissue with Congo Red. Detection of amyloid is relevant for a whole host of diseases, including Alzheimer’s disease, Creutzfeld-Jakob’s disease and Bovine Spongiform Encephalopathy. The birefringence of Congo Red-stained amyloid, appearing green under polarized light, improves detection relative to simple visible examination of the colored deposits. Additionally, upon binding to amyloid, Congo Red displays bright fluorescence emission at 614 nm. Wavelength Maxima: Excitation 497nm Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.