Learn More
Hach Company Radiometer Analytical XE100 Conductivity Cell (2-pole Pt sensor, k = 1.0 cm-1, glass body, screw cap)
Supplier:  Hach Company B60E100
Encompass_Preferred
The Radiometer XE100 is a 2-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1) and glass body. The XE100 sensor's 2 platinum plate design is ideal for routine conductivity measurements or for continuous measurement in various aqueous and non-aqueous media. The XE100 has a screw cap connection and requires the CL136 cable (part no. A94L136) for compatibility with Radiometer Analytical CDM210/230/92 conductivity meters or the CL143 cable (part no. A94L143) for compatibilty with Hach sensION+ benchtop conductivity meters.
Catalog No. 50-199-7542
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.