Learn More
Chemglass Life Sciences Condenser, Friedrichs, 330mm, 24/40 Joint, GL-14 Screw Caps, Removable Hose Connections
Supplier: Chemglass Life Sciences CG1210HC01
Condenser, Friedrichs, 330mm, 24/40 Joint, GL-14 Screw Caps, Removable Hose Connections. Friedrichs condenser similar to CG-1210 but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Hose connections have an O.D. of 8mm and an I.D. of 4mm. Supplied complete with two GL-14 open top screw caps. See CG-198 and CG-195 for replacement GL fittings.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.