Learn More
Chemglass Life Sciences Condenser, Cold Finger, 14/20 Inner Joint, 100mm Length Below Joint
Supplier: Chemglass Life Sciences CG122214
Condenser, Cold Finger, 14/20 Inner Joint, 100mm Length Below Joint. Condenser has an inner joint and a sealed lower drip tip for drop control. Hose connections have an O.D. of 10mm at the middle serration