Learn More
STEMCELL Technologies PBS-MINI 0.5 MAG Single-Use Vessel, Size: 4 Units
SDP
Supplier:  STEMCELL Technologies 1001007
Encompass
Reliably and rapidly scale up your 3D cell cultures and suspensions with the PBS-MINI MagDrive Bioreactor Base Unit and MAG Single-Use Vessels. The gentle yet efficient mixing provided by the Vertical-Wheel™ impeller enables the expansion of shear-sensitive cells without anti-foaming agents or shear protectants. Ideal for human pluripotent stem cells (hPSCs), the compact, sealed Base Unit can be used inside incubators. Conveniently control your culture system with a speed dial and digital display, and visualize cells in low-light conditions using built-in LED lights. This Base Unit is designed to be used with the 0.1 L and 0.5 L PBS-MINI MAG Single-Use Vessels, which it magnetically drives via MagDrive technology. These animal component-free disposable vessels are sold separately in packs of 4. To learn more about the PBS-MINI Vertical-Wheel™ Bioreactor, visit www.stemcell.com/pbs-mini. This product is for Research Use Only.
Catalog No. 50-225-0338
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.