Learn More
MilliporeSigma™ Supelco™ Column for Flash-Chromatography Assembly with Threaded Joints and Fritted Disc (Porosity C), Joint Size: ST/NS 45/50
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Specifications
Column Type Flash Chromatography
For Use With (Application) Flash Chromatography
Joint Size ST/NS 45/50
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity & Availability  
11-100-8749
View Documents
MilliporeSigma™ Supelco™
Z4166221EA
2000 mL 76 mm 560 mm N/A N/A N/A
Description
Description
Specifications
Specifications
Flash Chromatography
Flash Chromatography
ST/NS 45/50
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.