Learn More
Electron Microscopy Sciences Collodion (Parlodion), 2% in Amyl Acetate 30 ML
Supplier: Electron Microscopy Sciences 1262050
Highly purified nitrocellulose (parlodion strip) in glass distilled amyl acetate. Useful for forming a negative replica to very fine detail. Two types are available: Our sterile formula which is filtered down to 0.45 micron and our non-sterile formula. Sterile formula is filtered to 0.45 micron. 30 ml avoids hazardous shipping.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.