Learn More
CTL Scientific Supply Corp COBALT TEST PAPER,100 STRIPS
Supplier: CTL Scientific Supply Corp 90728
COBALT TEST PAPER - box of 100 strips (each strip 20 x 70 mm). The white test paper turns blue upon application of cobalt ions (Co 2+ ). Limit of sensitivity: 25 mg/l Co 2+. Note: Non-destructive tests for the detection of cobalt in alloys are carried out differently. Limit of sensitivity: 0.5 %
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.