Learn More
World Precision Instrument CDOM Coastal Ocean Water System, High Volume
SDP
Supplier:  World Precision Instrument CDOMCOASTHVB
Encompass_Preferred
What is CDOM?
Colored Dissolved Organic Matter (CDOM) is organic matter whose optical properties are measurable using WPI’s Liquid Waveguide Capillary Cell (LWCC). CDOM occurs naturally in water systems and is derived from organic tannins. CDOM concentration depends on the location where samples are taken, with coastal waters showing higher CDOM concentrations compared to open-ocean waters. In addition, CDOM absorption depends on open-ocean water depth. LWCCs are fiber optic flow cells that combine an increased optical pathlength (1–500 cm) with small sample volumes ranging from 2.4 μL to about 3 mL.
Catalog No. 50-252-1208
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.