Learn More
Enzo Life Sciences CNQX . disodium salt (5mg). CAS: 115066-14-3 (non-salt)
SDP
Supplier:  Enzo Life Sciences ALX550283M005
Encompass_Preferred
Water soluble, potent competitive kainate/quisqualate (non-NMDA) receptor antagonist. More soluble form of CNQX (ALX-550-042). Alternative name: 6-Cyano-7-nitroquinoxaline-2,3-dione . 2Na. Purity: ≥98% (TLC). Solubility: Soluble in water (5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9522
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.