Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clostridium Perfringens Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10062474JF549
Description
Clostridium Perfringens Polyclonal antibody specifically detects Clostridium Perfringens in Bacteria samples. It is validated for ELISASpecifications
Clostridium Perfringens | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Bacteria | |
Purified |
ELISA | |
Janelia Fluor 549 | |
C. perfringens | |
Clostridium perfringens ATCC 13124 | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction