Learn More
MilliporeSigma™ Supelco™ Click-On Inline Super Clean™ Gas Purifiers
Simple installation and replacement of the Click-On Purifier eliminates the risk of damage to gas line connections caused by overtightening
Supplier:  MilliporeSigma™ Supelco™ 28863U
 View more versions of this product
Catalog No. 11-101-6549
Description
Description
  • Simply attach the connectors to the gas line, install the purifier, and hand tighten the connectors
  • Click-on Connectors are not included and must be purchased before initial use
Specifications
Specifications
194 mm
1 Ea.
32 mm
Stainless Steel
Hydrocarbon Trap
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.