Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CLEC10A/CD301, Biotin, Clone: 121A5.01, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0011B100
Description
CLEC10A/CD301 Monoclonal specifically detects CLEC10A/CD301 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
CLEC10A/CD301 | |
Monoclonal | |
0.5mg/mL | |
Immunoprecipitation 1:10-1:500 | |
CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human | |
IgG1 |
Immunoprecipitation | |
121A5.01 | |
Biotin | |
NP_001754 | |
CLEC10A | |
CD301 was used as immunogen. | |
0.1 mg | |
Immunology | |
10462 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction