Learn More
Cleatech SDPVC 32" VERTICAL FLOW HOOD
Supplier: Cleatech 10007DFL
Portable Bench top Vertical Laminar Flow station, Work space 31.5"Wx 21.75"Dx 23.75"H , Clear Static Dissipative PVC construction, Clear Satic Dissipative PVC front tilt-up sash,110VAC, 50/60Hz. Adjustable speed Impeller, Backward Curve, 730 CFM @ 0.000-In,Including HEPA Filter , Including Built-in Fluorescent Light
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.