Learn More
TrueCare Biomedix CLEAN ROOM GOWN,STRL,40GSM,XL
Supplier: TrueCare Biomedix TCBA40STXL
Sterile Clean Room Gown, 40gsm+-3gsm White SMS,anti-static treated, stand up collar with velcro closure in the back, with elastic wrists, 1pcsterile bag,50pcsctn, XL
This item is not returnable.
View return policy