Learn More
TrueCare Biomedix CLEAN ROOM GOWN,STRL,40GSM,S
Supplier:  TrueCare Biomedix TCBA40STS
Encompass_Preferred
Sterile Clean Room Gown, 40gsm+-3gsm White SMS,anti-static treated, stand up collar with velcro closure in the back, with elastic wrists, 1pcsterile bag,50pcsctn, S
Catalog No. NC2217183
Due to product restrictions, please to purchase or view availability for this product.
Explore all special offers