Learn More
Electron Microscopy Sciences Aurion Cold Water Fish Skin Gelatine 40% 10 ML
Supplier: Electron Microscopy Sciences 25560
The use of Cold Water Fish Skin Gelatin to prevent background reactions has been recommended by e.g. Behnke et al. (J. Cell Biol. 41, [1986], 386). The product is supplied as a liquid concentrate (40%).
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Share a Content Correction