Learn More
Coast CL40R Rechargeable Clamp Light
The CL40R Clamp Light, the larger of our two clamp lights, packs bold power into precision task lighting—with an impressive 3900 lumens from its Wide Angle Flood Beam.
Supplier: Coast 30685
Description
- Featuring a high-strength aluminum clamp and bigger light body, this rock-solid work light attaches firmly to a wide variety of supports—from framing studs to the truck door
- The clamp also doubles as a dynamic stand when rested on a flat surface
- Once in place, the light rotates up to 360 degrees and tilts up to 115 degrees for the ultimate in lighting control
- Engineered with the COAST Dual Power system, the rechargeable CL40R runs on a built-in COAST ZITHION™ battery. In addition to the ZITHION™ Rechargeable Battery, the CL40R can also be continuously run using standard AC/DC power. To continuously run the Clamp Light, plug the USB-C cable (included) into the Clamp Light and plug the cable into an available power source, then turn the light on. While in a continuous run, the CL40R will operate at a single setting at a reduced level. The clamp light will not charge while in continuous run mode and must be turned off to fully charge. A battery life indicator ensures you know when more power is needed, and the battery also doubles as a power bank for external devices
- Constructed of top-grade materials, the CL40R is rated IP54 for water resistance.
Specifications
LED Clip Lamp | |
LED |