Learn More
Thermo Scientific™ Chromacol™ GOLD-Grade Inert Vials and Inserts
Ensure sample security and integrity with inert vials that have the highest recovery rates, an ultra-low pH shift, and lowest alkali content.
Supplier: Thermo Scientific™ 2SVG
Description
Maximize your sample performance and have fewer failed assays with Thermo Scientific™ Chromacol™ GOLD-Grade Inert Glass Vials. These vials offer the most state-of-the-art technology in the industry for the analysis of critical low detection limit analytes, and when sample quality, reproducibility, security, and integrity are of upmost importance.
For easier sample handling and less risk of sample loss, the Chromacol GOLD-Grade Vials are the best-in-class choice for all chromatography users performing ultra-trace analysis.
Choose the right vial for your needs- Get maximum sample extraction without need for separate inserts by using Microsampling and High Recovery Vials
- Identify sample and volume with graduated, write-on patch provided on certain vials
- Obtain maximum recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional)
- Polyspring inserts are self-aligning and provide a cushion against needle contact
- Precision point insert minimizes residual sample loss
- Pulled point inserts are an economical choice for noncritical applications
- Thermo Scientific™ Accela™
- Thermo Scientific™ Surveyor LC autosampler
- Beckman™
- CTC™
- Gilson™
- Knauer™
- Shimadzu™
- Spark™
- Varian™
- VWR™ (Merck)/Hitachi™
*See the Thermo Scientific Columns and Consumables Catalog for autosampler compatibility.
Specifications
2mL Screw Thread Vial, Flat Bottom, GOLD Grade Glass, Clear | |
12 mm | |
2.0 mL | |
Chromacol Gold | |
500 | |
No |
Clear | |
Glass | |
<170 μL | |
32 mm | |
8 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.