Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Chromacol™ GOLD-Grade Inert Vials and Inserts

Ensure sample security and integrity with inert vials that have the highest recovery rates, an ultra-low pH shift, and lowest alkali content.

Supplier:  Thermo Scientific™ 02MTVWG

 View more versions of this product

Catalog No. 13-622-255


Description

Description

Maximize your sample performance and have fewer failed assays with Thermo Scientific™ Chromacol™ GOLD-Grade Inert Glass Vials. These vials offer the most state-of-the-art technology in the industry for the analysis of critical low detection limit analytes, and when sample quality, reproducibility, security, and integrity are of upmost importance.

For easier sample handling and less risk of sample loss, the Chromacol GOLD-Grade Vials are the best-in-class choice for all chromatography users performing ultra-trace analysis.

Achieve confidence in results
  • Made of 1st hydrolytic class, special 33 expansion glass; meeting current USP requirements for pharmacopeias.
  • Ultra-low adsorption surface for critical polar compounds, enabling ultra trace-level analysis for strongly adsorbing analytes.
  • The high-class standard in Europe and Asia for decades.
  • Ultra-low pH shift due to the minimized surface activity.
  • Lowest alkaline content for less glass-wall interactions.
Increase productivity and save time
  • Highest recovery rates, even with critical compounds (e.g., trisubstituted N-atoms, tertiary amines) in trace concentrations.
  • Peaks show the same heights and symmetry and feature high recovery and reproducibility.
Reduce cost of analysis
  • Stable analysis and higher reproducibility means increased security and integrity of analytical results.
  • Reduced amount of buffer saves money.
  • No risk of run-stops during overnight autosampler runs.
  • Lot-independent, optimal autosampler fit.
Specifications

Specifications

0.2mL Insert, Pulled Point, GOLD Grade Glass, Clear
6 mm
200 μL
Chromacol Gold
1000
Clear
Glass
<4 μL
30 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.