Learn More
Eurofins Discoverx PathHunter CHO-K1 OPRL1 β-Arrestin Cell Line, 2 vials. (93-0264C2)
Supplier: Eurofins Discoverx 930264C2
The PathHunter OPRL1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. orphanin FQ) based activation of OPRL1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged OPRL1 and EA-tagged β-Arrestin-2. Activation of OPRL1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.