Learn More
Bioassay Systems CHLORIDE ASSAY KIT
SDP
Supplier:  Bioassay Systems DICL250
Encompass_Preferred
Assay kit, chloride ion and evaluation of drug effects on chloride metabolism. Method: OD610nm. Samples: biological, food and environment. Species: all. Procedure: 5 min. Size: 250 tests. Detection limit: 0.7 mg/dL (0.2 mM).
Catalog No. 50-489-212
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.