Learn More
*AVS BIO CHICK NEG SERUM LIQUID
Supplier: *AVS BIO 10100507
Sterile chicken serum negative for pathogens as per European Pharmacopeia. AllAntisera are heat inactivated.Size: 5 mi liquid
This item is not returnable.
View return policy