Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Taped Seams, Hood and Boot
Catalog No. 17184109 Shop All International Enviroguard Systems Products
Click to view available options
Size:
These durable and disposable coveralls with elastic wrist, ankle, and back protect against chemical splashes and toxic particles.
  • Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
  • A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
  • A high-contrast red stripe lets you quickly distinguish this taped-seam garment from coveralls made with serged seams
  • For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
  • With an attached hood and boots, you're protected from head to toe
  • Taped seams offer added protection over serged seam coveralls
  • A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
  • Elastic back stretches with the wearer for more comfort and better tear resistance
  • Elastic wrists and ankles offer a secure fit to keep sleeves and pant lets in place to prevent skin exposure
  • Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
Product Type
Chemical Protective Clothing
Size
3X-Large
Gender
Unisex
Material
Laminated Non-woven Fabric
Color
Yellow
Disposable
Single-use
Waist Style
Elastic at Back
Ankle Style
Attached Boots
Cuff Style
Elastic
Neck Style
Hooded
Hood Type
Attached, Elastic Face
Closure Type
2-way Front Zipper with Storm Flap
Seam Type
Serged, Taped
Sterility
Non-sterile
Resistance
Acid, Caustics, Chemicals, Isopropyl Alcohol, Particle
Latex Free
Yes
No. of Pockets
None