Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Coveralls with Taped Seams, Elastic Wrist and Ankle
Catalog No. 17184138 Shop All International Enviroguard Systems Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
These durable and disposable coveralls with taped seams and elastic back protect against chemical splashes and toxic particles.
  • Proprietary, hybrid membrane fabric offers a high moisture vapor transfer rate (MVTR), so you stay cool as you put in a hard day's work
  • A storm flap over the zipper with pressure-sensitive tape helps prevent hazardous liquids from seeping through exposed zipper seams
  • A high-contrast red stripe lets you quickly distinguish this taped-seam garment from coveralls made with serged seams
  • For added comfort, seamless shoulders help prevent tears and allow you to move more freely as you work
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure area
  • A wider, gusseted crotch gives the wearer more room to move in all directions without ripping fabric
  • Elastic back stretches with user for more comfort and better tear resistance
  • Storm flap over zipper with pressure-sensitive tape adds an extra layer of protection
  • High moisture vapor transfer rate improves wearer comfort
  • Elastic wrists offer a secure fit to keep sleeves in place to prevent skin exposure
  • Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
Product Type Chemical Protective Clothing
Size 2X-Large
Gender Unisex
Material Laminated Non-woven Fabric
Color Yellow
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Serged, Taped
Sterility Non-sterile
Resistance Acid, Caustics, Chemicals, Isopropyl Alcohol, Particle
Latex Free Yes
No. of Pockets None
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.