Learn More
Enzo Life Sciences Chemiluminescent labeling kit (1x5Reactions)
Supplier: Enzo Life Sciences ADI907001
Chemiluminescent acridinium esters are commonly used to label antibodies, proteins, nucleic acids, and peptides containing primary amine groups. These chemiluminescent molecules are frequently for use as detection reagents in immunoassays as a safe alternative to radioactive tracers. The Chemiluminescent labeling kit provides the necessary components for quick, efficient, and easy acridinium labeling. Applications: Chemiluminescence. Contents: Acridinium NHS Ester, Dry DMF, Bicarbonate labeling buffer, 10% Lysine, Gel filtration column, Column buffer concentrate, Set of Trigger solutions
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.