Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Chemical, Biohazard & Hazardous Material Label - Self-Sticking Polyester
Self-Sticking Polyester: Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.
Supplier: Brady™ 87764
Description
- Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
- Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
- Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
- Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Label | |
Permanent Acrylic | |
Globally Harmonized System of Classification and Labeling of Chemicals (GHS); NFPA 704; OSHA 1910.1200 | |
Label | |
5 x 3.5 in. (W x H) | |
Polyester | |
3.5 x 5 in. | |
Gloss | |
One |
Not Autoclavable | |
Black/Red on White | |
FLAMMABLE GAS | |
3.5 in. | |
English | |
1 Ea. | |
Self-adhesive | |
5 in. | |
127 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction