Learn More
Electron Microscopy Sciences Retriever Thermal Slide Processor Chamber Rack
Supplier: Electron Microscopy Sciences 6270502
Used with the Retriever benchtop thermal slide processer that is used on formalin-fixed, paraffin embedded tissues prior to immunostaining.