Learn More
Bevco Precision Manufacturing Co Doral Tall Height ESD/ISO 5 Cleanroom Blue Vinyl Chair w/Medium Back Upgrade; Articulating Seat & Back Tilt; Chrome Tubular Steel Base w/Welded Footring; Casters
Supplier:  Bevco Precision Manufacturing Co 5611E2BLVMDBCARD5S
Encompass_Preferred
BEVCO’s Doral ESD/ISO 5 Cleanroom Series models are designed to meet specific particulate goals of ISO 14644-1 and the ESD requirements of ANSI/ESD STM12.1-2019 delivering a combination ESD/Cleanroom seating solution for highly technical environments. Specifically, users that have a petite-to-average build will truly enjoy this seating experience.This articulating tilt model features simultaneous seat and back adjustments to properly position the angle of the chair for increased comfort during extended sitting durations. Fully adjustable medium back has a durable scuff resistant back pan to protect the upholstery.A waterfall seat promotes lower body circulation, improving overall wellness.The pneumatic height adjustment allows you to easily change your seat height from 26" - 31".Upholstered ESD vinyl meets CAL 117 standards.The chair features a sturdy 22" diameter chrome tubular steel base with brass drag chain and welded footring.Model comes with ESD hard floor casters.
Catalog No. NC2181988
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.