Learn More
Enzo Life Sciences Ceritinib (free base) (100mg). CAS: 1032900-25-6
Supplier: Enzo Life Sciences ENZCHM2180100
Ceritinib is a selective inhibitor of ALK, a target found in metastatic non-small cell lung cancer. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.