Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ceresin Wax, Spectrum™ Chemical
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Ceresin Wax , also known as ceresin or cerin, is a wax that comes from ozokerite through a purifying process. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
CAS Max % | 100% |
Packaging | Fiber Drum |
Quantity | 50 lb. |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Share a Content Correction