Learn More
STEMCELL Technologies TMRE (Perchlorate), Size: 25 mg
SDP
Supplier:  STEMCELL Technologies 1000992
Encompass_Preferred
Detect changes in mitochondrial membrane with TMRE (Perchlorate) dye. TMRE uses a quick and simple staining protocol that is well suited for high-throughput screening and is compatible with flow cytometry. This cell-permeable, cationic compound enables the distinction between polarized and depolarized cells. TMRE readily accumulates in mitochondria with the high mitochondrial membrane potential and emits a fluorescent red-orange signal with an emission of 574 nm. Depolarized mitochondria, however, fail to retain the dye and fluoresce. For more information on TMRE (Perchlorate), visit www.stemcell.com.
Catalog No. 50-225-0346
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.