Learn More
Bioassay Systems CELL VIABILITY KIT 5000 TESTS
SDP
Supplier:  Bioassay Systems CQBL05K
Encompass_Preferred
Assay kit, cell viability, proliferation, cytotoxcity and high-throughput screen for anticancer agents. Method: FL530/590nm. Samples: cell culture. Species: all. Procedure: 1-5 hrs. Size: 5000 tests. Detection limit: 100 cells.
Catalog No. 50-489-189
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.