Learn More
Mascon Medical Cell Strainer, Bulk Pack, 100m. Fits into nearly all 50ml conical tubes. 1EA/Bag, 50EA, Case.
SDP
Supplier:  Mascon Medical FSSTRAINER
Encompass_Preferred
Mascon Medical™ Cell Strainers, a more efficient alternative to gauze filtration, are perfect for stem cell and tissue-derived primary cell preparation. They ensure a consistently uniform single-cell suspension while the nylon mesh optimizes performance in a variety if applications. This cell strainers 100?m pores are evenly spaced to ensure consistent results and will fit into nearly all 50ml conical tubes. The extended lip on this strainer enables aseptic handling with forceps. These sterile cell strainers are Non-Pyrogenic and DNase/RNase free.Specifications: Volume: 100?m Qty. / Case: 50 Color: Yellow/Blue Sterile: Yes, by E-bean, SAL=10-6 Packaging Format: 1EA/Bag, 50EA/CaseFeatures: Non-Pyrogenic DNase/RNase free Fit nearly all 50ml conical tubes
Catalog No. NC2180272
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.