Learn More
Electron Microscopy Sciences ReadyStrain™ Cell Straining Kit, 40 µm, Blue
Supplier: Electron Microscopy Sciences 6474906
ReadyStrain provides all the necessary parts for filtration of cell suspensions and combines them into a single, preassembled, ready to use, sterile packet. These kits eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes strainers and caps).
The individual sterile packets mean researchers are no longer forced to expose entire bags of 50ml tubes and corresponding caps in order to perform one filtration. Instead they can open a single ReadyStrain packet directly under the hood.
Each kit contains a sterile SureStrain™ Strainer, pre-attached to a sterile 50ml tube, with a separate sterile screw-cap ready for assembly, processing and storage after the filtration is complete. Kits are supplied in easy open "tear-strip" bags and gamma sterilized.
Size: 40 µm
Color: Blue
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.