Learn More
Research Products International Corp Cell Spreaders, L Shaped, S, Individually Wrapped, 500/cs
SDP
Supplier:  Research Products International Corp 247662A
Encompass_Preferred
These cell spreaders have an L shape that provides quick and uniform cell distribution across surface of growth media. Spreaders are made from durable polystyrene for easy disposal after each use.
Catalog No. 50-223-0561
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.