Learn More
Bellco Glass, Inc. Cell Scraper 39cm 30mm
Supplier: Bellco Glass, Inc. 229315000
Cell Scrapers are designed to gently harvest cells from tissue culture flasks dishes or bottles. Soft pliable blade won’t damage cells or scratch surfaces. Scraper blade is positioned parallel to the handle for windshield wiper like scraping. Lifter blade is positioned perpendicular to the handle for use with a raking motion. Pivoting blade Cell Scraper features free rotating blade that adjusts to the desired position providing access to all corners of the flask or tissue culture vessel.Specifications Handle Length 39cm Blade Length 30mm Blade Orientation Scraper Parallel Case Quantity 100
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.