Learn More
Enzo Life Sciences Cell lysis buffer 4, (100 ml) (100ml)
SDP
Supplier:  Enzo Life Sciences ADI801339
Encompass_Preferred
Formulation: Liquid. 100mM PBS, pH 7.3, containing 150mM sodium chloride, 1mM EDTA, 0.5% Triton X-100 and 0.005% Tween 20. Long Term Storage: +4°C.
Catalog No. 50-200-6816
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.