Learn More
Hach Company Radiometer Analytical CDC641T-6 Conductivity Cell with Temperature Sensor (2-pole Pt sensor, k = 0.85 cm⁻¹, glass body)
Supplier:  Hach Company E61M022
Encompass_Preferred
The Radiometer CDC641T-6 is a 2-pole conductivity cell with platinized, platinum sensor (k = 0.85 cm-1), glass body and integrated temperature sensor. The CDC641T-6 sensor's 2 platinum plate design is ideal for routine conductivity measurements or for continuous measurement in various aqueous and non-aqueous media. Additionally, the CDC641T-6 meets requirements of EP 2.2.38. The cell has a fixed cable with RJ45 plug for use with Hach sensION+ benchtop conductivity meters.
Catalog No. 50-199-7617
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.