Learn More
Hach Company Radiometer Analytical CDC566T Conductivity Cell with Temperature Sensor (4-pole Pt sensor, k = 1.0 cm-1, epoxy body)
Supplier:  Hach Company E61M010
Encompass_Preferred
The Radiometer CDC566T Conductivity Cell is a 4-pole conductivity cell with non-platinized, platinum sensor (k = 1.0 cm-1), robust epoxy body and integrated temperature sensor. The CDC566T sensor's 4-pole platinum ring design ensures results in general aqueous and non-aqueous media over several decades of conductivity with a single cell, using just one calibration. The epoxy body of the probe can be easily removed for rinsing, which makes the cell ideal for measurements in harsh conditions. The CDC566T has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters. Please reference article no. E61M021 for the CDC536T (3-pole), compatible for use with Hach sensION+ benchtop conductivity meters.
Catalog No. 50-199-7611
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.