Learn More
Hach Company Radiometer Analytical CDC511T Conductivity Cell with Temperature Sensor (4-pole Pt sensor, k = 1.0 cm-1)
Supplier: Hach Company E61M009
The Radiometer CDC511T is a 4-pole conductivity cell with non-platinized, platinum sensor (k = 1.0 cm-1), integrated temperature sensor and TPX body. The CDC511T sensor's 4 platinum ring design ensures results in general aqueous and non-aqueous media over several decades of conductivity with a single cell, using just one calibration. The CDC511T can be used with a flow-through cell for pure water measurements and meets requirements of USP 24-NF19. It has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.