Learn More
Hach Company Radiometer Analytical CDC241 Conductivity Cell (2-pole Pt sensor, k = 1.0 cm-1, glass body, screw cap)
Supplier:  Hach Company E61M008
Encompass_Preferred
The Radiometer CDC241 is a 2-pole conductivity cell with non-platinized, platinum sensor (k = 1.0 cm-1) and glass body. The CDC241 sensor's 2 platinum plate design is ideal for continuous measurement or titration in viscous media. The CDC241 has a screw cap connection and requires the CL136 cable (part no. A94L136) for compatibility with Radiometer Analytical CDM210/230/92 conductivity meters or the CL143 cable (part no. A94L143) for compatibilty with Hach sensION+ benchtop conductivity meters.
Catalog No. 50-199-7609
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.