Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD69 Hamster anti-Mouse, BUV395, Clone: H1.2F3, BD Horizon™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 569367
Description
CD69 Monoclonal antibody specifically detects CD69 in Mouse samples. It is validated for Flow cytometrySpecifications
CD69 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
12515 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 λ3 |
Flow Cytometry | |
H1.2F3 | |
Brilliant Ultraviolet 395 | |
VEA; Very Early Activation Antigen; AIM; Activation Induced Molecule | |
Mouse Dendritic Epidermal T Cell Line Y245 | |
50 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction