Learn More
Fluidigm Corp CD56
SDP
Supplier:  Fluidigm Corp 3163007B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD56, also known as NCAM (neural cell adhesion molecule), Leu-19, and NKH1, is a type I transmembrane glycoprotein. CD56 is expressed on natural killer (NK) cells and NK-T cells as well as a host of neural lineages. It has also been reported to be expressed in some cancers including leukemia, small-cell lung carcinomas, neuronal tumors, and myelomas.
Catalog No. NC0859269