Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD54 Mouse anti-Human, Porcine, FITC, Clone: 15.2, Southern Biotech™
Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 944102
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD54 | |
Monoclonal | |
Lot specific | |
Supplied in PBS/NaN3 | |
Rheumatoid synovial cells and human monocytes | |
RUO | |
Human, Porcine | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
15.2 | |
FITC | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-8°C; Protect from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction