Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD49d Mouse anti-Human, Unconjugated, Clone: 7.2, Southern Biotech™
Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 943201
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD49d | |
Monoclonal | |
0.1mg/mL | |
Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added | |
0.1 mg | |
Primary | |
Store at 2-8°C | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
7.2 | |
Unconjugated | |
Mouse | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction