Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD40/TNFRSF5, Alexa Fluor 488, Clone: mAb89, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0131A488100
Description
CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunocytochemistry.Specifications
CD40/TNFRSF5 | |
mAb89 | |
Alexa Fluor 488 | |
PBS and 50% Glycerol with No Preservative | |
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
Mouse | |
IgG1 | |
0.1 mg | |
Adaptive Immunity, Apoptosis, Asthma, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
Primary | |
958 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
0.5mg/mL | |
Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence, Immunocytochemistry | |
Ion Exchange Chromatography | |
CD40 | |
Anti-u activated tonsillar human B cells. | |
Ion exchange chromatography | |
RUO | |
Store at -20C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction